Novus Biologicals
Manufacturer Code:NBP275585
Catalog # NBP275585
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JU41-80 |
Immunogen | Recombinant protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: Cytoplasmic protein NCK1; cytoplasmic protein NCK1 MGC12668 NCK adaptor protein 1melanoma NCK protein nck-1 NCKalpha NCKNCK tyrosine kinase non-catalytic region of tyrosine kinase SH2/SH3 adaptor protein NCK-alpha; melanoma NCK protein; NCK adaptor protein 1; NCK tyrosine kinase; Nck-1; non-catalytic region of tyrosine kinase; SH2/SH3 adaptor protein NCK-alpha
Gene Aliases: NCK; nck-1; NCK1; NCKalpha
UniProt ID: (Human) P16333
Entrez Gene ID: (Human) 4690
Molecular Function:
G-protein modulator
enzyme modulator
guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.