Abnova
Manufacturer Code:MAB16226
Catalog # 89-998-958
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 11D10 |
Immunogen | Recombinant His fusion protein corresponding to human NCK1 from <i>E. coli</i>. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Cytoplasmic protein NCK1; melanoma NCK protein; MGC12668; NCK; NCK adaptor protein 1; NCK tyrosine kinase; Nck-1; NCKalpha; non-catalytic region of tyrosine kinase; SH2/SH3 adaptor protein NCK-alpha
Gene Aliases: NCK; nck-1; NCK1; NCKalpha
UniProt ID: (Human) P16333
Entrez Gene ID: (Human) 4690
Molecular Function:
G-protein modulator
enzyme modulator
guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.