Abnova
Manufacturer Code:H00008440B01
Catalog # 89-006-563
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | NCK2 (NP_001004720 1 a.a. ~ 380 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human NCK2 protein.
For Research Use Only
Protein Aliases: Cytoplasmic protein NCK2; GRB4; Growth factor receptor-bound protein 4; NCK adaptor protein 2; Nck-2; NCKbeta; noncatalytic region of tyrosine kinase, beta; SH2/SH3 adaptor protein NCK-beta
Gene Aliases: GRB4; NCK2; NCKbeta
UniProt ID: (Human) O43639
Entrez Gene ID: (Human) 8440
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.