Invitrogen
Manufacturer Code:PA575191
Catalog # PIPA575191
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 115-170 of Human NCF4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: NCF-4; Neutrophil cytosol factor 4; Neutrophil cytosol factor 4 NCF-4 Neutrophil NADPH oxidase factor 4 SH3 and PX domain-containing protein 4 p40phox NCF4 SH3PXD4; neutrophil cytosolic factor 4, 40kDa; Neutrophil NADPH oxidase factor 4; p40-phox; p40phox; SH3 and PX domain-containing protein 4
Gene Aliases: AI451400; CGD3; NCF; NCF4; P40PHOX; SH3PXD4
UniProt ID: (Human) Q15080, (Mouse) P97369
Entrez Gene ID: (Human) 4689, (Rat) 500904, (Mouse) 17972
Molecular Function:
G-protein modulator
enzyme modulator
guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.