Invitrogen
Manufacturer Code:PA529000
Catalog # PIPA529000
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 1 and 298 of Human NCF4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: NCF P40PHOX SH3PXD4 NCF-4 SH3 and PX domain-containing protein 4 neutrophil NADPH oxidase factor 4 neutrophil cytosol factor 4 p40-phox; NCF-4; Neutrophil cytosol factor 4; neutrophil cytosolic factor 4, 40kDa; Neutrophil NADPH oxidase factor 4; p40-phox; p40phox; SH3 and PX domain-containing protein 4
Gene Aliases: CGD3; NCF; NCF4; P40PHOX; SH3PXD4
UniProt ID: (Human) Q15080
Entrez Gene ID: (Human) 4689
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.