Invitrogen
Manufacturer Code:PA514593
Catalog # PIPA514593
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 260-289 amino acids from the C-terminal region of human NCF4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: NCF-4; Neutrophil cytosol factor 4; Neutrophil cytosol factor 4 NCF-4 Neutrophil NADPH oxidase factor 4 p40-phox p40phox SH3 and PX domain-containing protein 4 SH3PXD4 NCF4; neutrophil cytosolic factor 4, 40kDa; Neutrophil NADPH oxidase factor 4; p40-phox; p40phox; SH3 and PX domain-containing protein 4
Gene Aliases: CGD3; NCF; NCF4; P40PHOX; SH3PXD4
UniProt ID: (Human) Q15080
Entrez Gene ID: (Human) 4689
Molecular Function:
G-protein modulator
enzyme modulator
guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.