Invitrogen
Manufacturer Code:MA534783
Catalog # PIMA534783
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JG86-31 |
Immunogen | Recombinant protein within Human NOXA2 aa 150-300. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles store in dark |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 67 kDa neutrophil oxidase factor; 67 kDa neutrophil oxidase factor FLJ93058 NADPH oxidase activator 2 Ncf2 NCF-2 Neutrophil cytosol factor 2 Neutrophil NADPH oxidase factor 2 Noxa2 P67PHOX p67-phox; chronic granulomatous disease, autosomal 2; NADPH oxidase activator 2; NCF-2; Neutrophil cytosol factor 2; neutrophil cytosolic factor 2 (65kD, chronic granulomatous disease, autosomal 2); Neutrophil NADPH oxidase factor 2; p67-phox
Gene Aliases: NCF-2; NCF2; NOXA2; P67-PHOX; P67PHOX
UniProt ID: (Human) P19878
Entrez Gene ID: (Human) 4688
Molecular Function:
oxidase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.