Invitrogen
Manufacturer Code:MA535393
Catalog # PIMA535393
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0828 |
Immunogen | A synthesized peptide derived from human NOXA2/p67phox |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 67 kDa neutrophil oxidase factor; 67 kDa neutrophil oxidase factor FLJ93058 NADPH oxidase activator 2 Ncf2 NCF-2 Neutrophil cytosol factor 2 Neutrophil NADPH oxidase factor 2 Noxa2 P67PHOX p67-phox; NADPH oxidase activator 2; NADPH oxidase subunit (67 kD); NADPH oxidase subunit (67kDa); NCF-2; ncf2; ncf2 {ECO:0000312|EMBL:FAA00361.1}; Neutrophil cytosol factor 2; Neutrophil NADPH oxidase factor 2; p67-phox
Gene Aliases: Ncf-2; Ncf2; NOXA2; P67phox
UniProt ID: (Mouse) O70145, (Rat) A7E3N2
Entrez Gene ID: (Mouse) 17970, (Rat) 364018
Molecular Function:
oxidase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.