Invitrogen
Manufacturer Code:PA552244
Catalog # PIPA552244
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human NCF2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 67 a neutrophil oxidase factor Chronic granulomatous disease autosomal 2 FLJ93058 NADPH oxidase activator 2 NCF-2 Ncf2 NCF2 Neutrophil cytosol factor 2 Neutrophil NADPH oxidase factor 2 NOXA2 p67-phox p67phox; 67 kDa neutrophil oxidase factor; chronic granulomatous disease, autosomal 2; NADPH oxidase activator 2; NCF-2; Neutrophil cytosol factor 2; neutrophil cytosolic factor 2 (65kD, chronic granulomatous disease, autosomal 2); Neutrophil NADPH oxidase factor 2; p67-phox
Gene Aliases: NCF-2; NCF2; NOXA2; P67-PHOX; P67PHOX
UniProt ID: (Human) P19878
Entrez Gene ID: (Human) 4688
Molecular Function:
oxidase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.