Abnova
Manufacturer Code:PAB29250
Catalog # 89-130-687
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S345 of human NCF1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human NCF1.
For Research Use Only
Protein Aliases: 47 kDa autosomal chronic granulomatous disease protein; 47 kDa neutrophil oxidase factor; FLJ79451; NADPH oxidase organizer 2; NCF-1; NCF-47K; NCF1A; Neutrophil cytosol factor 1; neutrophil cytosolic factor 1, (chronic granulomatous disease, autosomal 1); Neutrophil NADPH oxidase factor 1; Nox organizer 2; Nox-organizing protein 2; NOXO2; p47-phox; p47phox; SH3 and PX domain-containing protein 1A; SH3PXD1A
Gene Aliases: NCF1; NCF1A; NOXO2; p47phox; SH3PXD1A
UniProt ID: (Human) P14598
Entrez Gene ID: (Human) 653361
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.