Abnova
Manufacturer Code:PAB27626
Catalog # 89-129-137
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to internal region of mouse Ncf1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of Ncf1.
For Research Use Only
Protein Aliases: 47 kDa neutrophil oxidase factor; NADPH oxidase subunit (47 kD); NADPH oxidase subunit (47kDa); NCF-1; NCF-47K; Neutrophil cytosol factor 1; Neutrophil NADPH oxidase factor 1; NOXO2; p47; p47-phox; p47phox
Gene Aliases: Ncf-1; NCF-47K; Ncf1; NOXO2; p47-phox; p47<phox>; p47phox
UniProt ID: (Mouse) Q09014
Entrez Gene ID: (Mouse) 17969
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.