Invitrogen
Manufacturer Code:PA5114796
Catalog # PIPA5114796
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human AADACL1 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Aadacl1 Arylacetamide deacetylase-like 1 B230106I24Rik Chlorpyrifos oxon-binding protein CPO-BP Kiaa1363 NCEH Nceh1 Neutral cholesterol ester hydrolase 1 RGD1311104; Arylacetamide deacetylase-like 1; Chlorpyrifos oxon-binding protein; CPO-BP; NCEH; Neutral cholesterol ester hydrolase 1
Gene Aliases: AADACL1; B230106I24Rik; CPO-BP; KIAA1363; NCEH; NCEH1; RGD1311104
UniProt ID: (Human) Q6PIU2, (Mouse) Q8BLF1, (Rat) B2GV54
Entrez Gene ID: (Human) 57552, (Mouse) 320024, (Rat) 294930
Molecular Function:
hydrolase
lipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.