Novus Biologicals
Manufacturer Code:NBP241222
Catalog # NBP241222
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a 17 amino acid peptide near the center of human NCBP1. The immunogen is located within amino acids 420 - 470 of NCBP1. Amino Acid Squence: LSQDPESPKPKFVREV |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 80 kDa nuclear cap-binding protein; CBP80; CBP80nuclear cap binding protein subunit 1 80kD NCBP 80 kDa subunit NCBPMGC2087 nuclear cap binding protein subunit 1 80kDa nuclear cap-binding protein subunit 180 kDa nuclear cap-binding protein Sto1; NCBP 80 kDa subunit; nuclear cap binding protein subunit 1, 80kD; nuclear cap binding protein subunit 1, 80kDa; Nuclear cap-binding protein subunit 1
Gene Aliases: CBP80; NCBP; NCBP1; Sto1
UniProt ID: (Human) Q09161
Entrez Gene ID: (Human) 4686
Molecular Function: RNA binding protein mRNA processing factor mRNA splicing factor nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.