Abnova
Manufacturer Code:PAB27446
Catalog # 89-128-957
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 17 amino acids at internal region of human NCBP1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of NCBP1.
For Research Use Only
Protein Aliases: 80 kDa nuclear cap-binding protein; CBP80; MGC2087; NCBP; NCBP 80 kDa subunit; nuclear cap binding protein subunit 1, 80kD; nuclear cap binding protein subunit 1, 80kDa; Nuclear cap-binding protein subunit 1
Gene Aliases: CBP80; NCBP; NCBP1; Sto1
UniProt ID: (Human) Q09161
Entrez Gene ID: (Human) 4686
Molecular Function:
RNA binding protein
mRNA processing factor
mRNA splicing factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.