Proteintech
Manufacturer Code:103491AP150UL
Catalog # 50-172-6234
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NCBP1 Fusion Protein Ag0496 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 80 kDa nuclear cap-binding protein; CBP80; CBP80 NCBP NCBP 80 kDa subunit NCBP1; NCBP 80 kDa subunit; nuclear cap binding protein subunit 1, 80kD; nuclear cap binding protein subunit 1, 80kDa; Nuclear cap-binding protein subunit 1
Gene Aliases: AU014645; AW538051; CBP80; NCBP; NCBP1; Sto1
UniProt ID: (Human) Q09161, (Mouse) Q3UYV9
Entrez Gene ID: (Human) 4686, (Mouse) 433702
Molecular Function: RNA binding protein mRNA processing factor mRNA splicing factor nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.