Invitrogen
Manufacturer Code:PA530467
Catalog # PIPA530467
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 233 of Human BRRN1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: barren homolog 1; Barren homolog protein 1; BRRN1 CAP-H HCAP-H XCAP-H homolog barren homolog 1 barren homolog protein 1 chromosome-associated protein H condensin complex subunit 2; Chromosome-associated protein H; Condensin complex subunit 2; hCAP-H; Non-SMC condensin I complex subunit H; non-SMC condensin I complex, subunit H; XCAP-H homolog
Gene Aliases: BRRN; BRRN1; CAP-H; CAPH; KIAA0074; NCAPH
UniProt ID: (Human) Q15003
Entrez Gene ID: (Human) 23397
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.