Novus Biologicals
Manufacturer Code:NBP297603R
Catalog # NB102762
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human NCAPH fragment. |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: barren (Drosophila) homolog barren homolog (Drosophila) barren homolog 1 barren homolog 1 (Drosophila) Barren homolog protein 1 BRRN BRRN1CAP-H CAPH Chromosome-associated protein H condensin complex subunit 2 hCAP-H KIAA0074 Non-SMC condensin I complex subunit H non-SMC condensin I complex subunit H XCAP-H homolog; barren homolog 1; Barren homolog protein 1; Chromosome-associated protein H; Condensin complex subunit 2; hCAP-H; Non-SMC condensin I complex subunit H; non-SMC condensin I complex, subunit H; XCAP-H homolog
Gene Aliases: BRRN; BRRN1; CAP-H; CAPH; KIAA0074; NCAPH
UniProt ID: (Human) Q15003
Entrez Gene ID: (Human) 23397
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.