Proteintech
Manufacturer Code:245631AP150UL
Catalog # 50-173-4424
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NCAPG Fusion Protein Ag19926 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CAPG CHCG Condensin complex subunit 3 Condensin subunit CAP G HCAP G Melanoma antigen NY MEL 3 NCAPG NY MEL 3 NYMEL3 XCAP G homolog; chromosome condensation protein G; Chromosome-associated protein G; Condensin complex subunit 3; Condensin subunit CAP-G; hCAP-G; Melanoma antigen NY-MEL-3; Non-SMC condensin I complex subunit G; non-SMC condensin I complex, subunit G; XCAP-G homolog
Gene Aliases: CAPG; CHCG; NCAPG; NY-MEL-3; NYMEL3; YCG1
UniProt ID: (Human) Q9BPX3
Entrez Gene ID: (Human) 64151
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.