Novus Biologicals
Manufacturer Code:NB1001573
Catalog # NB1001573
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 1450 and the C-terminus (residue 1507) of human Chromosome-Associated Protein D3 using the numbering given in entry NP_056076.1 (GeneID 23310). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CAPD3 CAP-D3 FLJ42888 hCAP-D3condensin-2 complex subunit D3 hcp-6 hHCP-6 KIAA0056MGC104671 Non-SMC condensin II complex subunit D3 non-SMC condensin II complex subunit D3; Condensin-2 complex subunit D3; hCAP-D3; Non-SMC condensin II complex subunit D3; non-SMC condensin II complex, subunit D3
Gene Aliases: CAP-D3; CAPD3; hCAP-D3; hcp-6; hHCP-6; KIAA0056; NCAPD3
UniProt ID: (Human) P42695
Entrez Gene ID: (Human) 23310
Molecular Function:
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.