Invitrogen
Manufacturer Code:PA557886
Catalog # PIPA557886
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human NCAPD2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: chromosome condensation related SMC associated protein 1; Chromosome condensation-related SMC-associated protein 1; Chromosome-associated protein D2; Condensin complex subunit 1; hCAP-D2; Non-SMC condensin I complex subunit D2; Non-SMC condensin I complex subunit D2 CAP-D2 CNAP1 KIAA0159 hCAP-D2; non-SMC condensin I complex, subunit D2; XCAP-D2 homolog
Gene Aliases: CAP-D2; CAPD2; CNAP1; hCAP-D2; KIAA0159; NCAPD2
UniProt ID: (Human) Q15021
Entrez Gene ID: (Human) 9918
Molecular Function:
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.