Invitrogen
Manufacturer Code:PA5110016
Catalog # PIPA5110016
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein of human NCAPD2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2810406C15Rik 2810465G24Rik CAPD2 CAP-D2 Chromosome condensation-related SMC-associated protein 1 Chromosome-associated protein D2 CNAP1 Condensin complex subunit 1 hCAP-D2 KIAA0159 mCAP-D2 mKIAA0159 NCAPD2 Non-SMC condensin I complex subunit D2 RGD1562596 XCAP-D2 homolog; chromosome condensation related SMC associated protein 1; Chromosome condensation-related SMC-associated protein 1; Chromosome-associated protein D2; Condensin complex subunit 1; hCAP-D2; mCAP-D2; Non-SMC condensin I complex subunit D2; non-SMC condensin I complex, subunit D2; XCAP-D2 homolog
Gene Aliases: 2810406C15Rik; 2810465G24Rik; CAP-D2; CAPD2; CNAP1; hCAP-D2; KIAA0159; mKIAA0159; NCAPD2; RGD1562596
UniProt ID: (Human) Q15021, (Mouse) Q8K2Z4
Entrez Gene ID: (Human) 9918, (Rat) 362438, (Mouse) 68298
Molecular Function:
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.