Invitrogen
Manufacturer Code:PA5112624
Catalog # PIPA5112624
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Neural cell adhesion molecule 2 protein (209-333AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: fasciclin II NCAM2 N-CAM-2 Ncam-2 NCAM21 Neural cell adhesion molecule 2 Neural cell adhesion molecule RB-8 Ocam OCAM-GPI olfactory axon cell adhesion molecule R4B12 R4B12 antigen Rncam; N-CAM-2; NCAM-2; Neural cell adhesion molecule 2
Gene Aliases: NCAM2; NCAM21
UniProt ID: (Human) O15394
Entrez Gene ID: (Human) 4685
Molecular Function:
cell adhesion molecule
cytokine receptor
defense/immunity protein
hydrolase
immunoglobulin receptor superfamily
immunoglobulin superfamily cell adhesion molecule
phosphatase
protein phosphatase
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.