Invitrogen
Manufacturer Code:PA5110015
Catalog # PIPA5110015
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 500-690 of human NCAM2 (NP_004531.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: N-CAM-2; NCAM-2; NCAM2 N-CAM-2 Ncam-2 NCAM21 Neural cell adhesion molecule 2 Neural cell adhesion molecule RB-8 Ocam OCAM-GPI R4B12 Rncam; Neural cell adhesion molecule 2
Gene Aliases: NCAM2; NCAM21
UniProt ID: (Human) O15394
Entrez Gene ID: (Human) 4685
Molecular Function: cell adhesion molecule cytokine receptor defense/immunity protein hydrolase immunoglobulin receptor superfamily immunoglobulin superfamily cell adhesion molecule phosphatase protein phosphatase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.