Abnova
Manufacturer Code:PAB0134
Catalog # 89-113-972
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids 442-685 of Mouse NCAM-2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C on dry atmosphere.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against mouse NCAM-2 (442 a.a.-685 a.a) partial recombinant protein.
For Research Use Only
Protein Aliases: N-CAM-2; Ncam-2; Neural cell adhesion molecule 2; Neural cell adhesion molecule RB-8; Ocam; olfactory axon cell adhesion molecule; R4B12; R4B12 antigen; RNCAM
Gene Aliases: Ncam-2; Ncam2; Ocam; RNCAM
UniProt ID: (Mouse) O35136
Entrez Gene ID: (Mouse) 17968
Molecular Function: cell adhesion molecule cytokine receptor defense/immunity protein hydrolase immunoglobulin receptor superfamily immunoglobulin superfamily cell adhesion molecule phosphatase protein phosphatase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.