Invitrogen
Manufacturer Code:702670
Catalog # 70-267-0
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 9H4L65 |
Immunogen | Protein corresponding to human NBS1 (aa400-aa754) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: AT-V1 AT-V2 ATV NBS NBS1 P95 Nijmegen breakage syndrome 1 (nibrin) cell cycle regulatory protein p95 p95 protein of the MRE11/RAD50 complex; Cell cycle regulatory protein p95; Nibrin; Nijmegen breakage syndrome 1 (nibrin); Nijmegen breakage syndrome protein 1; p95 protein of the MRE11/RAD50 complex
Gene Aliases: AT-V1; AT-V2; ATV; NBN; NBS; NBS1; P95
UniProt ID: (Human) O60934
Entrez Gene ID: (Human) 4683
Molecular Function:
DNA binding protein
damaged DNA-binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.