Invitrogen
Manufacturer Code:PA534384
Catalog # PIPA534384
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 15 amino acid synthetic peptide near the carboxy terminus of human NIBRIN |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cell cycle regulatory protein p95; Nibrin; NIBRIN Nijmegen breakage syndrome protein 1 NBN AT-V1 AT-V2 ATV NBS NBS1 P95 Cell cycle regulatory protein p95; Nijmegen breakage syndrome 1 (nibrin); Nijmegen breakage syndrome protein 1; Nijmegen breakage syndrome protein 1 homolog; p95 protein of the MRE11/RAD50 complex
Gene Aliases: AT-V1; AT-V2; ATV; NBN; NBS; NBS1; P95
UniProt ID: (Human) O60934, (Mouse) Q9R207
Entrez Gene ID: (Human) 4683, (Mouse) 27354
Molecular Function: DNA binding protein damaged DNA-binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.