Invitrogen
Manufacturer Code:PA5106284
Catalog # PIPA5106284
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human NBR1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1A13B 1A1-3B B-box protein Ca125 Cell migration-inducing gene 19 protein IAI3B KIAA0049 M17s2 Membrane component chromosome 17 surface marker 2 Membrane component chromosome 17 surface marker 2 homolog membrane component chromosome 17 surface marker 2 (ovarian carcinoma antigen CA125) MIG19 migration-inducing protein 19 mKIAA0049 Nbr1 NBR1 autophagy cargo receptor neighbor of Brca1 gene 1 Neighbor of BRCA1 gene 1 protein next to BRCA1 gene 1 protein next to the Brca1 ovarian carcinoma antigen CA125 Protein 1A1-3B RGD1311421; B-box protein; Cell migration-inducing gene 19 protein; Membrane component chromosome 17 surface marker 2; Membrane component chromosome 17 surface marker 2 homolog; membrane component, chromosome 17, surface marker 2 (ovarian carcinoma antigen CA125); migration-inducing protein 19; neighbor of BRCA1 gene 1; Neighbor of BRCA1 gene 1 protein; Next to BRCA1 gene 1 protein; next to the Brca1; ovarian carcinoma antigen CA125; Protein 1A1-3B
Gene Aliases: 1A1-3B; 1A13B; Ca125; IAI3B; KIAA0049; M17S2; MIG19; mKIAA0049; NBR1; RGD1311421
UniProt ID: (Human) Q14596, (Mouse) P97432, (Rat) Q501R9
Entrez Gene ID: (Human) 4077, (Mouse) 17966, (Rat) 303554
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.