Abnova
Manufacturer Code:H00004077A01
Catalog # 89-104-860
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 2 a.a. &bsim 96 a.a) partial recombinant protein with GST tag.,NBR1 (NP_005890 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant NBR1.
For Research Use Only
Protein Aliases: 1A1-3B; B-box protein; Cell migration-inducing gene 19 protein; KIAA0049; M17S2; Membrane component chromosome 17 surface marker 2; membrane component, chromosome 17, surface marker 2 (ovarian carcinoma antigen CA125); MIG19; migration-inducing protein 19; neighbor of BRCA1 gene 1; Neighbor of BRCA1 gene 1 protein; Next to BRCA1 gene 1 protein; Protein 1A1-3B
Gene Aliases: 1A1-3B; 1A13B; IAI3B; KIAA0049; M17S2; MIG19; NBR1
UniProt ID: (Human) Q14596
Entrez Gene ID: (Human) 4077
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.