OriGene
Manufacturer Code:TA801410
Catalog # 50-167-8827
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI8E7 |
Immunogen | Human recombit protein fragment corresponding to amino acids 183-460 of human NBN produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AT-V1 AT-V2 ATV NBS NBS1 P95; Cell cycle regulatory protein p95; Nibrin; Nijmegen breakage syndrome 1 (nibrin); Nijmegen breakage syndrome protein 1; p95 protein of the MRE11/RAD50 complex
Gene Aliases: AT-V1; AT-V2; ATV; NBN; NBS; NBS1; P95
UniProt ID: (Human) O60934
Entrez Gene ID: (Human) 4683
Molecular Function:
DNA binding protein
damaged DNA-binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.