Invitrogen
Manufacturer Code:PA538608
Catalog # PIPA538608
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the C-terminal region of human NBL1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DAN domain family member 1; differential screening-selected gene aberrant in neuroblastoma; neuroblastoma candidate region, suppression of tumorigenicity 1; neuroblastoma suppression of tumorigenicity 1 D1S1733E DAN DAND1 NB NO3 DAN domain family member 1 differential screening-selected gene aberrant in neuroblastoma neuroblastoma candidate region suppression of tumorigenicity 1 neuroblastoma; Neuroblastoma suppressor of tumorigenicity 1; Protein N03; Zinc finger protein DAN
Gene Aliases: D1S1733E; DAN; DAND1; NB; NBL1; NO3
UniProt ID: (Human) P41271
Entrez Gene ID: (Human) 4681
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.