Invitrogen
Manufacturer Code:PA5103967
Catalog # PIPA5103967
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human NAV1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 9530089B19 9930003A20Rik C230080M11Rik Kiaa1151 KIAA1213 LOC685707 mKIAA1151 NAV1 neuron navigator 1 neuron navigator-1 POMFIL3 pore membrane and/or filament interacting like protein 3 pore membrane and/or filament-interacting-like protein 3 similar to neuron navigator 1 STEERIN1 Steerin-1 unc-53 homolog 1 UNC53H1; Neuron navigator 1; neuron navigator-1; pore membrane and/or filament interacting like protein 3; Pore membrane and/or filament-interacting-like protein 3; Steerin-1; Steerin1; Unc-53 homolog 1; unc53H1
Gene Aliases: 9530089B19; 9930003A20Rik; C230080M11Rik; KIAA1151; KIAA1213; mKIAA1151; NAV1; POMFIL3; steerin-1; STEERIN1; UNC53H1
UniProt ID: (Human) Q8NEY1, (Mouse) Q8CH77
Entrez Gene ID: (Human) 89796, (Mouse) 215690
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.