Novus Biologicals
Manufacturer Code:NBP184082
Catalog # NBP184082
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:AKSFVKPPSLANLDKVNSNSLDLPSSSDTTHASKVPDLHATSSASGGPLPSCFTPSPAPILNINSASFSQGLELMSGFSVPKETRMYPKLSGLHRSMESLQMPMSLPSAFPSSTPVPTPPAPPAAPTEEETEELTWSGSP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ12560 FLJ14203 KIAA1151POMFIL3DKFZp781D0314 KIAA1213 MGC14961 neuron navigator 1 neuron navigator-1 pore membrane and/or filament interacting like protein 3 Pore membrane and/or filament-interacting-like protein 3 Steerin-1 STEERIN1 Unc-53 homolog 1 unc53H1; Neuron navigator 1; neuron navigator-1; pore membrane and/or filament interacting like protein 3; Pore membrane and/or filament-interacting-like protein 3; Steerin-1; Unc-53 homolog 1; unc53H1
Gene Aliases: KIAA1151; KIAA1213; NAV1; POMFIL3; STEERIN1; UNC53H1
UniProt ID: (Human) Q8NEY1
Entrez Gene ID: (Human) 89796
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.