Invitrogen
Manufacturer Code:PA537999
Catalog # PIPA537999
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-SVDSRFRGKGIAK from the internal region of mouse Nat8l (aa211-223) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Camello-like protein 3; N-acetylaspartate synthetase; N-acetyltransferase 8-like (GCN5-related, putative); N-acetyltransferase 8-like 1110038O08Rik Shati N-acetyltransferase 8-like protein NAA synthetase protein Shati; N-acetyltransferase 8-like protein; NAA synthetase; Protein Shati; Shati
Gene Aliases: 1110038O08Rik; CML3; NACED; NAT8-LIKE; NAT8L; Shati
UniProt ID: (Human) Q8N9F0, (Mouse) Q3UGX3
Entrez Gene ID: (Human) 339983, (Mouse) 269642
Molecular Function:
acetyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.