OriGene
Manufacturer Code:TA500544
Catalog # 50-167-1680
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5A8 |
Immunogen | Full-length protein expressed in 293T cell transfected with human NAT8 expression vector |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Acetyltransferase 2; ATase2; ATase2 CCNAT CML1 GLA Hcml1 TSC501 TSC510; Camello-like protein 1; CCNAT; Cysteinyl-conjugate N-acetyltransferase; kidney- and liver-specific gene product; N-acetyltransferase 8; N-acetyltransferase 8 (camello like); N-acetyltransferase 8 (GCN5-related, putative); NAT8; probable N-acetyltransferase 8
Gene Aliases: ATase2; CCNAT; CML1; GLA; Hcml1; NAT8; TSC501; TSC510
UniProt ID: (Human) Q9UHE5
Entrez Gene ID: (Human) 9027
Molecular Function:
acetyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.