Invitrogen
Manufacturer Code:PA576257
Catalog # PIPA576257
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human NAT8. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acetyltransferase 2; ATase2; camello-like 4; Camello-like protein 1; Camello-like protein 4; CCNAT; Cysteinyl-conjugate N-acetyltransferase; GLA CML1 CCNAT Hcml1 ATase2 TSC501 TSC510; kidney- and liver-specific gene product; N-acetyltransferase 8; N-acetyltransferase 8 (camello like); N-acetyltransferase 8 (GCN5-related, putative); NAT8; probable N-acetyltransferase 8; probable N-acetyltransferase CML4; putative N-acetyltransferase Camello 4
Gene Aliases: 0610037O16Rik; ATase2; CCNAT; CML1; Cml4; GLA; Hcml1; NAT8; TSC501; TSC510
UniProt ID: (Human) Q9UHE5, (Mouse) Q9JIY7, (Rat) Q9QXT3
Entrez Gene ID: (Human) 9027, (Mouse) 68396, (Rat) 64570
Molecular Function:
acetyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.