Invitrogen
Manufacturer Code:PA567353
Catalog # PIPA567353
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human NAT8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acetyltransferase 2; ATase2; Camello-like protein 1; CCNAT; CML1 GLA Hcml1 TSC501 TSC510 N-acetyltransferase 8 (camello like) camello-like protein 1 kidney- and liver-specific gene product probable N-acetyltransferase 8; Cysteinyl-conjugate N-acetyltransferase; kidney- and liver-specific gene product; N-acetyltransferase 8; N-acetyltransferase 8 (camello like); N-acetyltransferase 8 (GCN5-related, putative); NAT8; probable N-acetyltransferase 8
Gene Aliases: ATase2; CCNAT; CML1; GLA; Hcml1; NAT8; TSC501; TSC510
UniProt ID: (Human) Q9UHE5
Entrez Gene ID: (Human) 9027
Molecular Function: acetyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.