Proteintech
Manufacturer Code:158071AP150UL
Catalog # 50-173-0495
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NAT5 Fusion Protein Ag8541 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Methionine N-acetyltransferase; N acetyltransferase 5 NAA20 NAT3 NAT5 NatB complex subunit NAT5; N-acetyltransferase 3 homolog; N-acetyltransferase 5; N-acetyltransferase 5 (ARD1 homolog, S. cerevisiae); N-acetyltransferase 5 (GCN5-related, putative); N-acetyltransferase 5, ARD1 subunit (arrest-defective 1, S. cerevisiae, homolog); N-alpha-acetyltransferase 20; N-alpha-acetyltransferase 20, NatB catalytic subunit; N-terminal acetyltransferase B complex catalytic subunit NAA20; N-terminal acetyltransferase B complex catalytic subunit NAT5; N-terminal acetyltransferase complex ARD1 subunit; NatB catalytic subunit; NatB complex subunit NAT5
Gene Aliases: 1500004D14Rik; 2900026I01Rik; AU041458; D2Ertd186e; dJ1002M8.1; NAA20; NAT3; NAT3P; NAT5; NAT5P
UniProt ID: (Human) P61599, (Mouse) Q9H0Y4
Entrez Gene ID: (Human) 51126, (Mouse) 67877, (Rat) 362228
Molecular Function:
acetyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.