Invitrogen
Manufacturer Code:PA569545
Catalog # PIPA569545
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N-terminal region of rat NAT15 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GNAT acetyltransferase; GNAT acetytransferase; HAT4; HAT4 NAT15; Histone acetyltransferase type B protein 4; hNaa60; N-acetyltransferase 15; N-acetyltransferase 15 (GCN5-related, putative); N-alpha-acetyltransferase 60; N-alpha-acetyltransferase F; NatF; natF catalytic subunit
Gene Aliases: 1200013P24Rik; AI315146; HAT4; NAA60; NAT15; NatF; RGD1308915; UNQ2771/PRO7155
UniProt ID: (Human) Q9H7X0, (Bovine) Q17QK9, (Mouse) Q9DBU2, (Rat) Q3MHC1
Entrez Gene ID: (Goat) 102187019, (Rabbit) 100339358, (Human) 79903, (Bovine) 514154, (Horse) 100065966, (Dog) 609456, (Mouse) 74763, (Rat) 363545
Molecular Function: acetyltransferase acyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.