Invitrogen
Manufacturer Code:PA523792
Catalog # PIPA523792
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 5-33 amino acids from the N-terminal region of human NAT14 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: K562 cell-derived leucine-zipper-like protein 1; K562 cells-derived leucine zipper-like protein 1; K562 cells-derived leucine-zipper-like protein 1; KLP1 K562 cell-derived leucine-zipper-like protein 1 K562 cells-derived leucine zipper-like protein 1 K562 cells-derived leucine-zipper-like protein 1 N-acetyltransferase 14; N-acetyltransferase 14; N-acetyltransferase 14 (GCN5-related, putative)
Gene Aliases: BC047219; KLP1; NAT14
UniProt ID: (Human) Q8WUY8, (Mouse) Q8BVG8
Entrez Gene ID: (Human) 57106, (Mouse) 269854
Molecular Function:
acetyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.