Invitrogen
Manufacturer Code:PA5115617
Catalog # PIPA5115617
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human NAT14 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BC047219 K562 cell-derived leucine-zipper-like protein 1 K562 cells-derived leucine zipper-like protein 1 K562 cells-derived leucine-zipper-like protein 1 KLP1 N-acetyltransferase 14 N-acetyltransferase 14 (GCN5-related putative) N-acetyltransferase 14 (putative) Nat14; K562 cell-derived leucine-zipper-like protein 1; K562 cells-derived leucine zipper-like protein 1; K562 cells-derived leucine-zipper-like protein 1; N-acetyltransferase 14; N-acetyltransferase 14 (GCN5-related, putative); zinc finger protein 628
Gene Aliases: BC047219; KLP1; NAT14; RGD1565991; Zfp628
UniProt ID: (Human) Q8WUY8, (Mouse) Q8BVG8
Entrez Gene ID: (Human) 57106, (Mouse) 269854, (Rat) 361500
Molecular Function: acetyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.