Novus Biologicals
Manufacturer Code:NBP255519
Catalog # NBP255519
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:PSHLSVREMREDEKPLVLEMLKAGVKDTENRVALHALTRPPALLLLAAASSGL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.3.1 EC 2.3.1.- K562 cell-derived leucine-zipper-like protein 1 K562 cells-derived leucine zipper-like protein 1 KLP1K562 cells-derived leucine-zipper-like protein 1 N-acetyltransferase 14 N-acetyltransferase 14 (GCN5-related putative); K562 cell-derived leucine-zipper-like protein 1; K562 cells-derived leucine zipper-like protein 1; K562 cells-derived leucine-zipper-like protein 1; N-acetyltransferase 14; N-acetyltransferase 14 (GCN5-related, putative)
Gene Aliases: KLP1; NAT14
UniProt ID: (Human) Q8WUY8
Entrez Gene ID: (Human) 57106
Molecular Function: acetyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.