Novus Biologicals
Manufacturer Code:NBP29307620UL
Catalog # NB028046
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-169 of human NAA50 (NP_079422.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.3.1.- FLJ13194 hSAN Mak3 homolog (S. cerevisiae) N(alpha)-acetyltransferase 50 NatE catalytic subunit N-acetyltransferase 13 (GCN5-related) N-acetyltransferase 5 N-acetyltransferase san homolog NAT13San NAT5SAN NatE catalytic subunit separation anxiety; hNaa50p; hNAT5; hSAN; N-acetyltransferase 13; N-acetyltransferase 13 (GCN5-related); N-acetyltransferase 5; N-acetyltransferase san homolog; N-alpha-acetyltransferase 50; N-alpha-acetyltransferase 50, NatE catalytic subunit; N-epsilon-acetyltransferase 50; NatE catalytic subunit
Gene Aliases: MAK3; NAA50; NAT13; NAT13P; NAT5; NAT5P; SAN
UniProt ID: (Human) Q9GZZ1
Entrez Gene ID: (Human) 80218
Molecular Function: acetyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.