Proteintech
Manufacturer Code:161201AP150UL
Catalog # 50-173-0729
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NAT13 Fusion Protein Ag9075 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hNaa50p; hNAT5; hNAT5 hSAN MAK3 N acetyltransferase 13 N acetyltransferase 5 NAA50 NAT13 NAT5 SAN; hSAN; Mak3 homolog; Mak3p homolog; N-acetyltransferase 13; N-acetyltransferase 13 (GCN5-related); N-acetyltransferase 5; N-acetyltransferase NAT13; N-acetyltransferase san homolog; N-alpha-acetyltransferase 50; N-alpha-acetyltransferase 50, NatE catalytic subunit; N-epsilon-acetyltransferase 50; Nalpha acetyltransferase 50; NatE catalytic subunit
Gene Aliases: 2600005K24Rik; 2810441M03Rik; AW112078; MAK3; Mak3p; NAA50; NAT13; NAT13P; NAT5; NAT5P; SAN
UniProt ID: (Human) Q9GZZ1, (Mouse) Q6PGB6
Entrez Gene ID: (Human) 80218, (Rat) 288108, (Mouse) 72117
Molecular Function:
acetyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.