Invitrogen
Manufacturer Code:PA520882
Catalog # PIPA520882
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 15 amino acid peptide near the carboxy terminus of human NAT11. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: N(alpha)-acetyltransferase 40, NatD catalytic subunit, homolog; N-acetyltransferase 11; N-acetyltransferase 11 (GCN5-related, putative); N-alpha acetyl transferase 40; N-alpha-acetyltransferase 40; N-alpha-acetyltransferase 40, NatD catalytic subunit; N-alpha-acetyltransferase D; Nalpha acetyltransferase 40; NAT11 PATT1 N-acetyltransferase 11 (GCN5-related putative) N-alpha acetyl transferase 40 N-alpha-acetyltransferase 40 N-alpha-acetyltransferase 40 NatD catalytic subunit natD catalytic subunit; NatD; natD catalytic subunit; Protein acetyltransferase 1
Gene Aliases: 4931433E08Rik; AU023197; NAA40; NAT11; PATT1
UniProt ID: (Human) Q86UY6, (Mouse) Q8VE10
Entrez Gene ID: (Human) 79829, (Mouse) 70999
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.