Proteintech
Manufacturer Code:133651AP150UL
Catalog # 50-172-8682
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NAT10 Fusion Protein Ag4184 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 18S rRNA cytosine acetyltransferase; ALP KIAA1709 N acetyltransferase 10 NAT10 NET43; hALP; N-acetyltransferase 10; N-acetyltransferase 10 (GCN5-related); N-acetyltransferase-like protein; novel putative ATPase domain containing protein; RNA cytidine acetyltransferase
Gene Aliases: AI429152; ALP; KIAA1709; NAT10; NET43; RGD1306717
UniProt ID: (Human) Q9H0A0, (Mouse) Q8K224
Entrez Gene ID: (Human) 55226, (Mouse) 98956, (Rat) 311257
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.