Invitrogen
Manufacturer Code:PA576021
Catalog # PIPA576021
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human NAT1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Arylamide acetylase 1; Arylamine N-acetyltransferase 1; Arylamine N-acetyltransferase 1 Arylamide acetylase 1 Monomorphic arylamine N-acetyltransferase MNAT N-acetyltransferase type 1 NAT-1 NAT1 AAC1; MNAT; Monomorphic arylamine N-acetyltransferase; N(alpha)-acetyltransferase 15, NatA auxiliary subunit; N-acetyl transferase 1, liver, blood; N-acetyltransferase 1 (arylamine N-acetyltransferase); N-acetyltransferase type 1; NAT-1
Gene Aliases: AAC1; MNAT; NAT-1; NAT1; NATI
UniProt ID: (Human) P18440, (Mouse) P50294
Entrez Gene ID: (Human) 9, (Mouse) 17960
Molecular Function: acetyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.