Invitrogen
Manufacturer Code:PA524835
Catalog # PIPA524835
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 152-180 amino acids from the N-terminal region of human NASP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLB7527 PRO1999 histone H1-binding protein nuclear autoantigenic sperm protein; histone H1-binding protein; NASP; NASP histone chaperone; Nuclear autoantigenic sperm protein; nuclear autoantigenic sperm protein (histone-binding)
Gene Aliases: FLB7527; HMDRA1; NASP; PRO1999
UniProt ID: (Human) P49321
Entrez Gene ID: (Human) 4678
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.