Invitrogen
Manufacturer Code:PA5106317
Catalog # PIPA5106317
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human NASP corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5033430J04Rik AI131596 AI317140 D4Ertd767e DKFZp547F162 EMBL:AAH81913.1} Epcs32 FLB7527 FLJ31599 FLJ35510 histone H1-binding protein HMDRA1 MGC19722 MGC20372 MGC2297 Nasp nasp {ECO:0000312 NASP histone chaperone Nasp-T Nuclear autoantigenic sperm protein nuclear autoantigenic sperm protein (histone-binding) PRO1999 somatic histone binding protein NASP; histone H1-binding protein; NASP; NASP histone chaperone; nasp {ECO:0000312|EMBL:AAH81913.1}; Nuclear autoantigenic sperm protein; nuclear autoantigenic sperm protein (histone-binding); somatic histone binding protein NASP
Gene Aliases: 5033430J04Rik; AI131596; AI317140; D4Ertd767e; Epcs32; FLB7527; HMDRA1; NASP; Nasp-T; PRO1999
UniProt ID: (Human) P49321, (Rat) Q66HD3, (Mouse) Q99MD9
Entrez Gene ID: (Human) 4678, (Rat) 298441, (Mouse) 50927
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.