OriGene
Manufacturer Code:CF806499
Catalog # 50-166-7858
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI10E2 |
Immunogen | Full length human recombit protein of human NARS2 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AsnRS; asparagine tRNA ligase 2, mitochondrial (putative); Asparaginyl-tRNA synthetase; deafness, autosomal recessive 94; DFNB94 SLM5 asnRS; Probable asparagine--tRNA ligase, mitochondrial; probable asparaginyl-tRNA synthetase, mitochondrial
Gene Aliases: asnRS; DFNB94; NARS2; SLM5
UniProt ID: (Human) Q96I59
Entrez Gene ID: (Human) 79731
Molecular Function:
RNA binding protein
aminoacyl-tRNA synthetase
ligase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.