Novus Biologicals
Manufacturer Code:NBP272876AF594
Catalog # NP272876A94
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI10C10 |
Immunogen | Full length recombinant protein of human NARS2 (NP_078954) produced in HEK293T cells. |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: AsnRS; asnRS asparagine tRNA ligase 2 mitochondrial (putative) Asparagine--tRNA ligase asparaginyl-tRNA synthetase 2 mitochondrial (putative) EC 6.1.1 EC 6.1.1.22 FLJ23441 probable asparaginyl-tRNA synthetase mitochondrial SLM5; asparagine tRNA ligase 2, mitochondrial (putative); Asparaginyl-tRNA synthetase; deafness, autosomal recessive 94; Probable asparagine--tRNA ligase, mitochondrial; probable asparaginyl-tRNA synthetase, mitochondrial
Gene Aliases: asnRS; DFNB94; NARS2; SLM5
UniProt ID: (Human) Q96I59
Entrez Gene ID: (Human) 79731
Molecular Function: RNA binding protein aminoacyl-tRNA synthetase ligase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.